Assay Details
Gene Symbol: Slc27a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031736.1 | NP_113924.1 | 9 - 10 | 2478 | 62 | |
GenBank mRNA | BC081766.1 | - | 9 - 10 | 1815 | 62 | |
D85100.1 | - | 9 - 10 | 2478 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
very long-chain fatty acid metabolic process
fatty acid alpha-oxidation
long-chain fatty acid metabolic process
fatty acid beta-oxidation
bile acid biosynthetic process
fatty acid transport
very long-chain fatty acid catabolic process
long-chain fatty acid import into cell
methyl-branched fatty acid metabolic process
long-chain fatty acid-CoA ligase activity
signaling receptor binding
protein binding
ATP binding
fatty acid transmembrane transporter activity
enzyme binding
very long-chain fatty acid-CoA ligase activity
phytanate-CoA ligase activity
pristanate-CoA ligase activity
very long-chain fatty acid metabolic process
fatty acid alpha-oxidation
long-chain fatty acid metabolic process
fatty acid beta-oxidation
bile acid biosynthetic process
fatty acid transport
very long-chain fatty acid catabolic process
long-chain fatty acid import into cell
methyl-branched fatty acid metabolic process
long-chain fatty acid-CoA ligase activity
signaling receptor binding
protein binding
ATP binding
fatty acid transmembrane transporter activity
enzyme binding
very long-chain fatty acid-CoA ligase activity
phytanate-CoA ligase activity
pristanate-CoA ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P103632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101889 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12655 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12655 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P12655 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P997755 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12655 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700625164 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12197 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59245 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73283 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88959 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44340 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38130 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700458952 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700625164 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10839450 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839451 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839454 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839449 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839444 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839448 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839446 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839452 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839453 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839447 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839445 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10839443 | - | - |
Rat Genome 230 2.0 Array | 1368150_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368150_at | - | - |
Affymetrix HT Rat Focus Array | 1368150_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368150_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300011557.rn.1 | TC0300001642.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300011558.rn.1 | TC0300001642.rn.1 | RTA TC |
Rat Expression Set 230 | 1368150_PM_at | - | - |
Rat Expression Set 230 | 1368150_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839443 | - | - |
Rat Genome U34 Set | D85100_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22323350 | - | - |
