Assay Details
Gene Symbol: Arfgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033707.1 | NP_001028879.1 | 3 - 4 | 314 | 62 | |
XM_006234529.2 | XP_006234591.1 | 3 - 4 | 419 | 62 | ||
XM_006234530.2 | XP_006234592.1 | 3 - 4 | 418 | 62 | ||
XM_006234531.2 | XP_006234593.1 | 3 - 4 | 418 | 62 | ||
GenBank mRNA | BC101917.1 | - | 3 - 4 | 314 | 62 | |
FQ218185.1 | - | 3 - 4 | 386 | 62 | ||
FQ228393.1 | - | 3 - 4 | 381 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
