Assay Details
Gene Symbol: Rnf128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173349.1 | NP_001166820.1 | 1 - 2 | 679 | 70 | |
GenBank mRNA | BC167061.1 | - | 1 - 2 | 679 | 70 | |
FQ210020.1 | - | 1 - 2 | 667 | 70 | ||
FQ218340.1 | - | 1 - 2 | 667 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
