Assay Details
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762189.1 | XP_008760411.1 | 8 - 9 | 1691 | 69 | |
GenBank mRNA | FQ212553.1 | - | 4 - 5 | 476 | 69 |
Gene Symbol: Gabpb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135015.1 | NP_001128487.1 | 1 | 999 | 69 | |
XM_006254075.2 | XP_006254137.1 | 2 | 1032 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
