Assay Details
Gene Symbol: Smox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134854.1 | NP_001128326.1 | 4 - 5 | 1670 | 64 | |
XM_006235063.2 | XP_006235125.1 | 5 - 6 | 1562 | 64 | ||
XM_006235064.2 | XP_006235126.1 | 5 - 6 | 1561 | 64 | ||
XM_006235065.2 | XP_006235127.1 | 5 - 6 | 1561 | 64 | ||
XM_008762165.1 | XP_008760387.1 | 5 - 6 | 1578 | 64 | ||
GenBank mRNA | BC160836.1 | - | 5 - 6 | 1445 | 64 | |
BC161894.1 | - | 4 - 5 | 1670 | 64 |
Gene Symbol: RGD1564480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_577020.6 | XP_577020.4 | 1 | 1461 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
