Assay Details
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761954.1 | XP_008760176.1 | 3 - 4 | 579 | 110 | |
XM_008761955.1 | XP_008760177.1 | 3 - 4 | 468 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
