Assay Details
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761954.1 | XP_008760176.1 | 4 - 5 | 926 | 86 | |
XM_008761955.1 | XP_008760177.1 | 4 - 5 | 815 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
