Assay Details
Gene Symbol: Nr1h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031627.2 | NP_113815.2 | 3 - 4 | 511 | 62 | |
XM_006234533.2 | XP_006234595.1 | 4 - 5 | 689 | 62 | ||
XM_006234534.1 | XP_006234596.1 | 4 - 5 | 630 | 62 | ||
XM_006234535.2 | XP_006234597.1 | 4 - 5 | 831 | 62 | ||
GenBank mRNA | BC088750.1 | - | 3 - 4 | 511 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
