Assay Details
Gene Symbol: Trmt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107779.1 | NP_001101249.1 | 10 - 11 | 1413 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P015227 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P869351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165908 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P869351 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P015227 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P869351 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P015227 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P015227 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P869351 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P869351 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1047331 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38059 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10850169 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850168 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850162 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850163 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850166 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850161 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850167 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850164 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10850160 | - | - |
Rat Genome 230 2.0 Array | 1381217_at | - | - |
Rat Genome 230 2.0 Array | 1377625_at | - | - |
Rat Genome 230 2.0 Array | 1377021_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377021_at | - | - |
Rat Genome 230 2.0 Array | 1381217_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377625_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381217_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377021_at | - | - |
Affymetrix HT Rat Focus Array | 1377021_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377625_at | - | - |
Affymetrix HT Rat Focus Array | 1377625_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381217_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300032924.rn.1 | TC0300004228.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300032923.rn.1 | TC0300004228.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300032921.rn.1 | TC0300004228.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300032922.rn.1 | TC0300004228.rn.1 | RTA TC |
Rat Expression Set 230 | 1381217_PM_at | - | - |
Rat Expression Set 230 | 1377021_PM_at | - | - |
Rat Expression Set 230 | 1377021_at | - | - |
Rat Expression Set 230 | 1381217_at | - | - |
Rat Expression Set 230 | 1377625_PM_at | - | - |
Rat Expression Set 230 | 1377625_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850160 | - | - |
Rat Genome U34 Set | rc_AI639158_at | - | - |
Rat Genome U34 Set | AA944061_at | - | - |
Rat Genome U34 Set | rc_AI137337_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20916196 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354246 | - | - |
