Assay Details
Gene Symbol: Acp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016988.2 | NP_058684.2 | 6 - 7 | 678 | 94 | |
XM_006234500.1 | XP_006234562.1 | 7 - 8 | 1600 | 94 | ||
GenBank mRNA | BC081823.1 | - | 6 - 7 | 649 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
