Assay Details
Gene Symbol: Creb3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005562.1 | NP_001005562.1 | 10 - 11 | 1649 | 73 | |
XM_006234584.1 | XP_006234646.1 | 10 - 11 | 1705 | 73 | ||
GenBank mRNA | BC081951.1 | - | 10 - 11 | 1649 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
