Assay Details
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015032.3 | NP_001015032.2 | 6 - 7 | 1497 | 143 | |
XM_006234290.2 | XP_006234352.1 | 7 - 8 | 1732 | 143 | ||
GenBank mRNA | AB040674.1 | - | 6 - 7 | 1386 | 143 | |
BC101886.1 | - | 6 - 7 | 1500 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
