Assay Details
Gene Symbol: Phactr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764195.1 | XP_008762417.1 | 7 - 8 | 951 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
