Assay Details
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022502.2 | NP_071947.1 | 6 - 7 | 630 | 71 | |
GenBank mRNA | BC126089.1 | - | 6 - 7 | 630 | 71 | |
FQ213474.1 | - | 6 - 7 | 642 | 71 | ||
FQ213911.1 | - | 642 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
