Assay Details
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183056.2 | NP_898879.2 | 3 - 4 | 360 | 74 | |
XM_006238881.2 | XP_006238943.1 | 3 - 4 | 2070 | 74 | ||
GenBank mRNA | BC100062.1 | - | 3 - 4 | 360 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
