Assay Details
Gene Symbol: Gale | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080783.2 | NP_542961.2 | 10 - 11 | 1018 | 105 | |
XM_006239134.2 | XP_006239196.1 | 10 - 11 | 1983 | 105 | ||
XM_008764197.1 | XP_008762419.1 | 10 - 11 | 1249 | 105 | ||
GenBank mRNA | BC097293.1 | - | 10 - 11 | 1018 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
