Assay Details
Gene Symbol: Slc2a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764332.1 | XP_008762554.1 | 6 - 7 | 762 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
