Assay Details
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145766.2 | NP_665709.2 | 13 - 14 | 1511 | 57 | |
XM_006239527.2 | XP_006239589.1 | 12 - 13 | 2067 | 57 | ||
XM_006239528.2 | XP_006239590.1 | 13 - 14 | 1874 | 57 | ||
XM_006239529.2 | XP_006239591.1 | 13 - 14 | 1795 | 57 | ||
XM_006239530.1 | XP_006239592.1 | 13 - 14 | 1495 | 57 | ||
XM_006239531.2 | XP_006239593.1 | 13 - 14 | 1599 | 57 | ||
XM_006239532.2 | XP_006239594.1 | 8 - 9 | 1943 | 57 | ||
XM_006239533.1 | XP_006239595.1 | 8 - 9 | 920 | 57 | ||
GenBank mRNA | DY544650.1 | - | 1 - 2 | 107 | 57 | |
EX490221.1 | - | 5 - 6 | 601 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
