Assay Details
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145766.2 | NP_665709.2 | 4 - 5 | 399 | 74 | |
XM_006239527.2 | XP_006239589.1 | 3 - 4 | 955 | 74 | ||
XM_006239528.2 | XP_006239590.1 | 4 - 5 | 762 | 74 | ||
XM_006239529.2 | XP_006239591.1 | 4 - 5 | 683 | 74 | ||
XM_006239530.1 | XP_006239592.1 | 4 - 5 | 383 | 74 | ||
XM_006239531.2 | XP_006239593.1 | 4 - 5 | 487 | 74 | ||
GenBank mRNA | FM031851.1 | - | 4 - 5 | 399 | 74 | |
FM044847.1 | - | 4 - 5 | 370 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
