Assay Details
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145766.2 | NP_665709.2 | 6 - 7 | 677 | 67 | |
XM_006239527.2 | XP_006239589.1 | 5 - 6 | 1233 | 67 | ||
XM_006239528.2 | XP_006239590.1 | 6 - 7 | 1040 | 67 | ||
XM_006239529.2 | XP_006239591.1 | 6 - 7 | 961 | 67 | ||
XM_006239530.1 | XP_006239592.1 | 6 - 7 | 661 | 67 | ||
XM_006239531.2 | XP_006239593.1 | 6 - 7 | 765 | 67 | ||
GenBank mRNA | DY556124.1 | - | 1 - 2 | 97 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
