Assay Details
Gene Symbol: Hmgcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024386.1 | NP_077362.1 | 8 - 9 | 892 | 66 | |
GenBank mRNA | BC061797.1 | - | 8 - 9 | 894 | 66 | |
FQ209616.1 | - | 8 - 9 | 896 | 66 | ||
FQ234382.1 | - | 8 - 9 | 890 | 66 | ||
Y10054.1 | - | 8 - 9 | 892 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
