Assay Details
Gene Symbol: Ppie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047868.1 | NP_001041333.1 | 4 - 5 | 217 | 85 | |
XM_006238796.2 | XP_006238858.1 | 3 - 4 | 232 | 85 | ||
XM_006238797.2 | XP_006238859.1 | 3 - 4 | 241 | 85 | ||
GenBank mRNA | AY325152.1 | - | 4 - 5 | 217 | 85 | |
BC158560.1 | - | 3 - 4 | 182 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006238797.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
