Assay Details
Gene Symbol: Smap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100669.1 | NP_001094139.1 | 3 - 4 | 696 | 108 | |
GenBank mRNA | BC161927.1 | - | 3 - 4 | 690 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
