Assay Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146061.1 | NP_001139533.1 | 6 - 7 | 799 | 61 | |
NM_013214.4 | NP_037346.2 | 6 - 7 | 727 | 61 | ||
XM_006239447.2 | XP_006239509.1 | 6 - 7 | 827 | 61 | ||
XM_006239448.1 | XP_006239510.1 | 6 - 7 | 735 | 61 | ||
GenBank mRNA | BC087716.1 | - | 6 - 7 | 679 | 61 | |
D88891.1 | - | 6 - 7 | 799 | 61 | ||
Y09332.1 | - | 6 - 7 | 627 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
