Assay Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146061.1 | NP_001139533.1 | 7 - 8 | 919 | 78 | |
NM_013214.4 | NP_037346.2 | 7 - 8 | 847 | 78 | ||
XM_006239447.2 | XP_006239509.1 | 7 - 8 | 947 | 78 | ||
XM_006239448.1 | XP_006239510.1 | 7 - 8 | 855 | 78 | ||
GenBank mRNA | BC087716.1 | - | 7 - 8 | 799 | 78 | |
FQ212652.1 | - | 4 - 5 | 398 | 78 | ||
FQ231740.1 | - | 4 - 5 | 398 | 78 | ||
Y09332.1 | - | 7 - 8 | 747 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
