Assay Details
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100709.1 | NP_001094179.1 | 1 - 2 | 114 | 87 | |
XM_006238932.2 | XP_006238994.1 | 2 - 3 | 281 | 87 | ||
XM_006238933.2 | XP_006238995.1 | 2 - 3 | 243 | 87 | ||
XM_008764140.1 | XP_008762362.1 | 2 - 3 | 299 | 87 | ||
GenBank mRNA | BC160881.1 | - | 1 - 2 | 114 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
