Assay Details
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100709.1 | NP_001094179.1 | 7 - 8 | 789 | 61 | |
XM_006238932.2 | XP_006238994.1 | 8 - 9 | 956 | 61 | ||
XM_006238933.2 | XP_006238995.1 | 8 - 9 | 918 | 61 | ||
XM_008764140.1 | XP_008762362.1 | 8 - 9 | 974 | 61 | ||
GenBank mRNA | BC099218.1 | - | 3 - 4 | 393 | 61 | |
BC160881.1 | - | 7 - 8 | 789 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
