Assay Details
Gene Symbol: Tnfrsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019135.1 | NP_062008.1 | 11 - 12 | 1112 | 75 | |
GenBank mRNA | D42117.1 | - | 11 - 12 | 1112 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
