Assay Details
Gene Symbol: Park7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277249.1 | NP_001264178.1 | 4 - 5 | 367 | 91 | |
NM_001277250.1 | NP_001264179.1 | 4 - 5 | 367 | 91 | ||
NM_001277251.1 | NP_001264180.1 | 5 - 6 | 426 | 91 | ||
NM_001277252.1 | NP_001264181.1 | 4 - 5 | 307 | 91 | ||
NM_001277253.1 | NP_001264182.1 | 4 - 5 | 419 | 91 | ||
NM_057143.2 | NP_476484.1 | 4 - 5 | 549 | 91 | ||
GenBank mRNA | AF157511.1 | - | 4 - 5 | 549 | 91 | |
AF157512.1 | - | 4 - 5 | 303 | 91 | ||
AJ007291.1 | - | 5 - 6 | 474 | 91 | ||
BC091128.1 | - | 4 - 5 | 340 | 91 | ||
FM055257.1 | - | 4 - 5 | 337 | 91 | ||
FM061417.1 | - | 4 - 5 | 288 | 91 | ||
FM069664.1 | - | 4 - 5 | 358 | 91 | ||
FQ220895.1 | - | 4 - 5 | 419 | 91 | ||
FQ223564.1 | - | 4 - 5 | 417 | 91 | ||
FQ224480.1 | - | 5 - 6 | 410 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
