Assay Details
Gene Symbol: Rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107912.2 | NP_001101382.1 | 4 - 5 | 485 | 104 | |
GenBank mRNA | BC168994.1 | - | 4 - 5 | 485 | 104 |
Gene Symbol: LOC681419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037919.1 | 1 - 2 | 109 | 104 | ||
GenBank mRNA | EV762401.1 | - | 1 - 2 | 109 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
