Assay Details
Gene Symbol: Rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107912.2 | NP_001101382.1 | 6 - 7 | 772 | 77 | |
GenBank mRNA | BC168994.1 | - | 6 - 7 | 772 | 77 |
Gene Symbol: LOC681419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037919.1 | 3 - 4 | 396 | 77 | ||
GenBank mRNA | EV762401.1 | - | 3 - 4 | 396 | 77 |
Gene Symbol: RGD1563620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_227252.8 | XP_227252.5 | 2 | 759 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
