Assay Details
Gene Symbol: Rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107912.2 | NP_001101382.1 | 8 - 9 | 969 | 117 | |
GenBank mRNA | BC168994.1 | - | 8 - 9 | 969 | 117 |
Gene Symbol: LOC681419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037919.1 | 5 - 6 | 594 | 118 | ||
GenBank mRNA | EV762401.1 | - | 5 - 6 | 594 | 118 |
Gene Symbol: RGD1563620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_227252.8 | XP_227252.5 | 3 | 968 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
