Assay Details
Gene Symbol: Fam76a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108686.1 | NP_001102156.1 | 7 - 8 | 935 | 65 | |
GenBank mRNA | FQ211538.1 | - | 5 - 6 | 868 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P479251 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P222779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168110 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P479251 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P222779 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P222779 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P479251 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P222779 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P479251 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P222779 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57755 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10872634 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872636 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872637 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872638 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872629 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872632 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872631 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872630 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872635 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872633 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10872628 | - | - |
Rat Genome 230 2.0 Array | 1393610_at | - | - |
Rat Genome 230 2.0 Array | 1390171_at | - | - |
Rat Genome 230 2.0 Array | 1390171_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393610_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393610_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393610_at | - | - |
Rat Expression Set 230 | 1390171_PM_at | - | - |
Rat Expression Set 230 | 1390171_at | - | - |
Rat Expression Set 230 | 1393610_at | - | - |
Rat Expression Set 230 | 1393610_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872628 | - | - |
Rat Genome U34 Set | rc_AA924717_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21133864 | - | - |
