Assay Details
Gene Symbol: Casp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031632.1 | NP_113820.1 | 4 - 5 | 787 | 65 | |
GenBank mRNA | AF262319.1 | - | 4 - 5 | 787 | 65 | |
AF271996.1 | - | 4 - 5 | 754 | 65 | ||
AF286006.1 | - | 4 - 5 | 787 | 65 | ||
AY008275.1 | - | 4 - 5 | 754 | 65 | ||
AY027667.1 | - | 4 - 5 | 763 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
