Assay Details
Gene Symbol: Flvcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199109.2 | NP_954540.2 | 7 - 8 | 1683 | 63 | |
GenBank mRNA | AY260573.1 | - | 7 - 8 | 1435 | 63 | |
BC101899.1 | - | 7 - 8 | 1683 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
