Assay Details
Gene Symbol: Ahsa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115034.1 | NP_001108506.1 | 4 - 5 | 609 | 79 | |
GenBank mRNA | BC158698.1 | - | 4 - 5 | 566 | 79 | |
FQ223544.1 | - | 4 - 5 | 593 | 79 | ||
FQ232113.1 | - | 4 - 5 | 571 | 79 | ||
FQ233084.1 | - | 4 - 5 | 571 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
