Assay Details
Gene Symbol: Coq6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011983.1 | NP_001011983.1 | 4 - 5 | 536 | 76 | |
GenBank mRNA | BC079342.1 | - | 4 - 5 | 536 | 76 | |
FQ228932.1 | - | 3 - 4 | 893 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
