Assay Details
Gene Symbol: Coq6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011983.1 | NP_001011983.1 | 7 - 8 | 834 | 57 | |
GenBank mRNA | BC079342.1 | - | 7 - 8 | 834 | 57 | |
FQ228932.1 | - | 6 - 7 | 1191 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
