Assay Details
Gene Symbol: Smoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002835.1 | NP_001002835.1 | 9 - 10 | 937 | 76 | |
XM_006240284.2 | XP_006240346.1 | 9 - 10 | 1243 | 76 | ||
XM_006240285.2 | XP_006240347.1 | 9 - 10 | 1243 | 76 | ||
XM_008764785.1 | XP_008763007.1 | 10 - 11 | 1343 | 76 | ||
XM_008764786.1 | XP_008763008.1 | 10 - 11 | 1310 | 76 | ||
GenBank mRNA | FQ222878.1 | - | 9 - 10 | 1194 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
