Assay Details
Gene Symbol: Tc2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025152.1 | NP_001020323.1 | 6 - 7 | 751 | 84 | |
XM_008764831.1 | XP_008763053.1 | 5 - 6 | 2414 | 84 | ||
XM_008764832.1 | XP_008763054.1 | 6 - 7 | 2063 | 84 | ||
XM_008764833.1 | XP_008763055.1 | 6 - 7 | 2310 | 84 | ||
XM_008764834.1 | XP_008763056.1 | 5 - 6 | 1284 | 84 | ||
XM_008764835.1 | XP_008763057.1 | 6 - 7 | 1315 | 84 | ||
XM_008764836.1 | XP_008763058.1 | 6 - 7 | 933 | 84 | ||
XM_008764837.1 | XP_008763059.1 | 6 - 7 | 1278 | 84 | ||
XM_008764838.1 | XP_008763060.1 | 5 - 6 | 1247 | 84 | ||
XM_008764839.1 | XP_008763061.1 | 5 - 6 | 813 | 84 | ||
XM_008764840.1 | XP_008763062.1 | 4 - 5 | 680 | 84 | ||
GenBank mRNA | BC088432.1 | - | 6 - 7 | 751 | 84 | |
FQ211170.1 | - | 6 - 7 | 869 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
