Assay Details
Gene Symbol: Noxred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750205.3 | XP_003750253.1 | 6 - 7 | 1209 | 75 | |
XM_006240401.2 | XP_006240463.1 | 6 - 7 | 1025 | 75 | ||
XM_006240402.2 | XP_006240464.1 | 6 - 7 | 1222 | 75 | ||
XM_006240403.2 | XP_006240465.1 | 6 - 7 | 1355 | 75 | ||
XM_008764893.1 | XP_008763115.1 | 7 - 8 | 1106 | 75 | ||
XM_008764894.1 | XP_008763116.1 | 7 - 8 | 1320 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
