Assay Details
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013174.2 | NP_037306.1 | 6 - 7 | 1479 | 59 | |
XM_006240366.1 | XP_006240428.1 | 7 - 8 | 1823 | 59 | ||
GenBank mRNA | BC092195.1 | - | 6 - 7 | 1466 | 59 | |
U03491.1 | - | 6 - 7 | 1479 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
in utero embryonic development
transforming growth factor beta receptor signaling pathway
salivary gland morphogenesis
female pregnancy
aging
negative regulation of DNA replication
negative regulation of cell population proliferation
animal organ morphogenesis
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of macrophage cytokine production
cell growth
lung development
positive regulation of bone mineralization
negative regulation of transforming growth factor beta receptor signaling pathway
regulation of epithelial cell differentiation
mammary gland development
response to progesterone
positive regulation of collagen biosynthetic process
response to laminar fluid shear stress
wound healing
positive regulation of apoptotic process
negative regulation of neuron apoptotic process
response to estrogen
ossification involved in bone remodeling
cell-cell junction organization
regulation of endothelial cell differentiation
positive regulation of DNA replication
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
lung alveolus development
cell development
digestive tract development
embryonic neurocranium morphogenesis
inner ear development
regulation of epithelial cell proliferation
positive regulation of protein secretion
positive regulation of filopodium assembly
positive regulation of cell division
roof of mouth development
face morphogenesis
frontal suture morphogenesis
regulation of SMAD protein signal transduction
positive regulation of SMAD protein signal transduction
SMAD protein signal transduction
detection of hypoxia
cellular response to growth factor stimulus
type II transforming growth factor beta receptor binding
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
type I transforming growth factor beta receptor binding
type III transforming growth factor beta receptor binding
identical protein binding
protein heterodimerization activity
transforming growth factor beta binding
response to hypoxia
in utero embryonic development
transforming growth factor beta receptor signaling pathway
salivary gland morphogenesis
female pregnancy
aging
negative regulation of DNA replication
negative regulation of cell population proliferation
animal organ morphogenesis
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of macrophage cytokine production
cell growth
lung development
positive regulation of bone mineralization
negative regulation of transforming growth factor beta receptor signaling pathway
regulation of epithelial cell differentiation
mammary gland development
response to progesterone
positive regulation of collagen biosynthetic process
response to laminar fluid shear stress
wound healing
positive regulation of apoptotic process
negative regulation of neuron apoptotic process
response to estrogen
ossification involved in bone remodeling
cell-cell junction organization
regulation of endothelial cell differentiation
positive regulation of DNA replication
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
lung alveolus development
cell development
digestive tract development
embryonic neurocranium morphogenesis
inner ear development
regulation of epithelial cell proliferation
positive regulation of protein secretion
positive regulation of filopodium assembly
positive regulation of cell division
roof of mouth development
face morphogenesis
frontal suture morphogenesis
regulation of SMAD protein signal transduction
positive regulation of SMAD protein signal transduction
SMAD protein signal transduction
detection of hypoxia
cellular response to growth factor stimulus
type II transforming growth factor beta receptor binding
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
type I transforming growth factor beta receptor binding
type III transforming growth factor beta receptor binding
identical protein binding
protein heterodimerization activity
transforming growth factor beta binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P125025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7938 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P125025 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P125025 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P125025 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P125025 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701279026 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39711 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101146 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82513 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69457 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701279026 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10891305 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891304 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891306 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891307 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891308 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891309 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891310 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10891303 | - | - |
Rat Genome 230 2.0 Array | 1367859_at | - | - |
Rat Genome 230 2.0 Array | 1367859_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367859_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367859_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600020474.rn.1 | TC0600002883.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020475.rn.1 | TC0600002883.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600020476.rn.1 | TC0600002883.rn.1 | RTA TC |
Rat Expression Set 230 | 1367859_at | - | - |
Rat Expression Set 230 | 1367859_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10891303 | - | - |
Rat Genome U34 Set | U03491_g_at | - | - |
Rat Genome U34 Set | U03491_at | - | - |
Rat Genome U34 Set | rc_AA998464_at | - | - |
Rat Genome U34 Set | rc_AI008087_i_at | - | - |
Rat Genome U34 Set | U03491_at | - | - |
Rat Genome U34 Set | U03491_g_at | - | - |
Rat Genome U34 Set | U03491_at | - | - |
Rat Genome U34 Set | U03491_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22110808 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352414 | - | - |
RatRef-12 Expression BeadChip | ILMN_65300 | - | - |
