Assay Details
Gene Symbol: Alkbh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240394.2 | XP_006240456.1 | 4 - 5 | 565 | 80 | |
GenBank mRNA | FQ230972.1 | - | 2 - 3 | 140 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
