Assay Details
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013106.1 | NP_037238.1 | 4 - 5 | 541 | 97 | |
GenBank mRNA | DQ120473.1 | - | 4 - 5 | 434 | 97 | |
DQ120474.1 | - | 4 - 5 | 434 | 97 | ||
FQ222046.1 | - | 4 - 5 | 604 | 97 | ||
J03219.1 | - | 4 - 5 | 550 | 97 | ||
M20713.1 | - | 4 - 5 | 541 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
