Assay Details
Gene Symbol: Bdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106473.1 | NP_001099943.1 | 3 - 4 | 378 | 102 | |
XM_006233322.2 | XP_006233384.1 | 3 - 4 | 287 | 102 | ||
GenBank mRNA | BC095848.1 | - | 2 - 3 | 772 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
