Assay Details
Gene Symbol: Bdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106473.1 | NP_001099943.1 | 5 - 6 | 575 | 81 | |
XM_006233322.2 | XP_006233384.1 | 5 - 6 | 484 | 81 | ||
GenBank mRNA | BC095848.1 | - | 4 - 5 | 969 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
