Assay Details
Gene Symbol: Slc30a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191715.1 | NP_001178644.1 | 7 - 8 | 855 | 83 | |
GenBank mRNA | BC091417.1 | - | 3 - 4 | 213 | 83 | |
BC107441.1 | - | 3 - 4 | 214 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
