Assay Details
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025271.1 | NP_001020442.1 | 3 - 4 | 1367 | 67 | |
GenBank mRNA | BC098731.1 | - | 3 - 4 | 1367 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
