Assay Details
Gene Symbol: Ddost | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012104.1 | NP_001012104.1 | 2 - 3 | 268 | 109 | |
GenBank mRNA | BC082075.1 | - | 2 - 3 | 268 | 109 | |
FQ228905.1 | - | 2 - 3 | 300 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
