Assay Details
Gene Symbol: Masp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172043.1 | NP_742040.1 | 4 - 5 | 578 | 72 | |
XM_006239400.2 | XP_006239462.1 | 4 - 5 | 634 | 72 | ||
XM_008764285.1 | XP_008762507.1 | 4 - 5 | 634 | 72 | ||
GenBank mRNA | AJ277747.1 | - | 3 - 4 | 520 | 72 | |
BC128700.1 | - | 4 - 5 | 579 | 72 | ||
Y18285.1 | - | 2 - 3 | 283 | 72 | ||
Y18564.1 | - | 3 - 4 | 357 | 72 | ||
Y18566.1 | - | 1 - 2 | 110 | 72 | ||
Y18572.1 | - | 3 - 4 | 415 | 72 | ||
Y18573.1 | - | 3 - 4 | 550 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
